accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1707-03-5

OHPO

CAS RN 1707-03-5

IUPAC Name: diphenylphosphinic acid
Synonyms: diphenylphosphinic acid
Molecular Weight (g/mol): 218.19
Molecular Formula: C12H11O2P
InChi Key: BEQVQKJCLJBTKZ-UHFFFAOYSA-N
SMILES: OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)