accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2

CAS RN 17075-03-5

IUPAC Name: pyren-4-amine
Synonyms: pyren-4-amine
Molecular Weight (g/mol): 217.27
Molecular Formula: C16H11N
InChi Key: AYBSKADMMKVOIV-UHFFFAOYSA-N
SMILES: NC1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)