CAS RN 170853-04-0
IUPAC Name:
tert-butyl N-[(3-formylphenyl)methyl]carbamate
Synonyms:
tert-butyl N-[(3-formylphenyl)methyl]carbamate
Molecular Weight (g/mol):
235.28
Molecular Formula:
C13H17NO3
InChi Key:
JORXNWZTJLYRQA-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC1=CC=CC(C=O)=C1
1
–
1
of
1
results
1
tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™
CAS: 170853-04-0 Molecular Formula: C13H17NO3 Molecular Weight (g/mol): 235.283 InChI Key: JORXNWZTJLYRQA-UHFFFAOYSA-N PubChem CID: 2794834 IUPAC Name: tert-butyl N-[(3-formylphenyl)methyl]carbamate SMILES: CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C=O