accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 170961-15-6

H3CH3CCH3OONHNS

CAS RN 170961-15-6

IUPAC Name: tert-butyl N-(1,3-thiazol-2-yl)carbamate
Synonyms: tert-butyl N-(1,3-thiazol-2-yl)carbamate
Molecular Weight (g/mol): 200.26
Molecular Formula: C8H12N2O2S
InChi Key: NCJXQSNROJRSSL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)