accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOI

CAS RN 1711-02-0

IUPAC Name: 4-iodobenzoyl chloride
Synonyms: 4-iodobenzoyl chloride
Molecular Weight (g/mol): 266.46
Molecular Formula: C7H4ClIO
InChi Key: NJAKCIUOTIPYED-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)