accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOBr

CAS RN 1711-09-7

IUPAC Name: 3-bromobenzoyl chloride
Synonyms: 3-bromobenzoyl chloride
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: PBOOZQFGWNZNQE-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(Br)=C1
Boiling Point 
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)