accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3ONH3C

CAS RN 17147-42-1

IUPAC Name: ethyl 3,5-dimethyl-1,2-oxazole-4-carboxylate
Synonyms: ethyl 3,5-dimethyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 169.18
Molecular Formula: C8H11NO3
InChi Key: OQRHVDXUJAQVNT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)ON=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results