accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOOHO

CAS RN 17153-20-7

IUPAC Name: 3-methyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: YBLSBWHFPXDRHC-UHFFFAOYSA-N
SMILES: CC1=NOC=C1C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results