accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17180-94-8

NNCl

CAS RN 17180-94-8

IUPAC Name: 5-chloropyrimidine
Synonyms: pyrimidine, 5-chloro-
Molecular Weight (g/mol): 114.53
Molecular Formula: C4H3ClN2
InChi Key: ZLNPDTOTEVIMMY-UHFFFAOYSA-N
SMILES: ClC1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results