accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1719-71-7

IUPAC Name: 2-(naphthalen-1-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 2-(naphthalen-1-yl)-3,5-diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 384.87
Molecular Formula: C23H17ClN4
InChi Key: RONADMZTCCPLEF-UHFFFAOYSA-M
SMILES: [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)