accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1720-32-7

IUPAC Name: [(1E,3E,5E)-6-phenylhexa-1,3,5-trien-1-yl]benzene
Synonyms: 1,6-diphenyl-1,3,5-hexatriene diphenylhexatriene 1,6-diphenylhexatriene
Molecular Weight (g/mol): 232.33
Molecular Formula: C18H16
InChi Key: BOBLSBAZCVBABY-WPWUJOAOSA-N
SMILES: C(=C\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)