accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NONH2N

CAS RN 17220-38-1

IUPAC Name: 1,2,5-oxadiazole-3,4-diamine
Synonyms: furazandiamine
Molecular Weight (g/mol): 100.08
Molecular Formula: C2H4N4O
InChi Key: JHJVSUCUNFXIHN-UHFFFAOYSA-N
SMILES: NC1=NON=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results