accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHO

CAS RN 17238-05-0

IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: (+/-)-dihydrodaidzein 4',7-dihydroxy-isoflavanone dihydrodaidzein
Molecular Weight (g/mol): 256.26
Molecular Formula: C15H12O4
InChi Key: JHYXBPPMXZIHKG-UHFFFAOYNA-N
SMILES: OC1=CC=C(C=C1)C1COC2=CC(O)=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results