accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH

CAS RN 17264-02-7

IUPAC Name: 1-cyclohexylpropan-1-ol
Synonyms: 1-cyclohexylpropan-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: JVTXOMXEPFDMHB-UHFFFAOYNA-N
SMILES: CCC(O)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results