accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 172732-52-4

BOON

CAS RN 172732-52-4

IUPAC Name: 2-(1,3,2-dioxaborinan-2-yl)benzonitrile
Synonyms: 2-(1,3,2-dioxaborinan-2-yl)benzonitrile
Molecular Weight (g/mol): 187.01
Molecular Formula: C10H10BNO2
InChi Key: REQZFVYFYAZUMG-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1B1OCCCO1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)