accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOS

CAS RN 17277-58-6

IUPAC Name: methyl 2-(phenylsulfanyl)acetate
Synonyms: methyl 2-(phenylsulfanyl)acetate
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: MUNSXQQODXYRKI-UHFFFAOYSA-N
SMILES: COC(=O)CSC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)