accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OOHNN

CAS RN 173340-23-3

IUPAC Name: tert-butyl N-[(1-benzylpiperidin-4-yl)methyl]carbamate
Synonyms: tert-butyl N-[(1-benzylpiperidin-4-yl)methyl]carbamate
Molecular Weight (g/mol): 304.43
Molecular Formula: C18H28N2O2
InChi Key: SOXSDIKJHXTJEP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results