accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OCN

CAS RN 17337-13-2

IUPAC Name: 2-isocyanato-1,1'-biphenyl
Synonyms: 2-isocyanato-1,1'-biphenyl
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: IHHUGFJSEJSCGE-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)