accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(S)OHN

CAS RN 17342-08-4

IUPAC Name: (5S)-5-(hydroxymethyl)pyrrolidin-2-one
Synonyms: (5S)-5-(hydroxymethyl)pyrrolidin-2-one
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: HOBJEFOCIRXQKH-BYPYZUCNSA-N
SMILES: OC[C@@H]1CCC(=O)N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)