CAS RN 17345-61-8
IUPAC Name:
3,4-dihydroxybenzonitrile
Synonyms:
3,4-dihydroxybenzonitrile
Molecular Weight (g/mol):
135.12
Molecular Formula:
C7H5NO2
InChi Key:
NUWHYWYSMAPBHK-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1O)C#N
1
–
1
of
1
results
1
3,4-Dihydroxybenzonitrile, 97%, Thermo Scientific Chemicals
CAS: 17345-61-8 Molecular Formula: C7H5NO2 Molecular Weight (g/mol): 135.122 MDL Number: MFCD00016436 InChI Key: NUWHYWYSMAPBHK-UHFFFAOYSA-N Synonym: protocatachuonitrile,3,4-dihydroxy benzonitrile,3,4-dihydroxybenzenecarbonitrile,protocatechuonitrile,4-cyanocatechol,protocatechunitrile,4-cyanopyrocatechol,pubchem3120,acmc-209e6q,3, 4-dihydroxybenzonitrile PubChem CID: 87065 IUPAC Name: 3,4-dihydroxybenzonitrile SMILES: C1=CC(=C(C=C1C#N)O)O