accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17348-69-5

ONONCl

CAS RN 17348-69-5

IUPAC Name: 5-chloro-2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 5-chloro-2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 170.55
Molecular Formula: C6H3ClN2O2
InChi Key: DHPQXIQZZCNOLI-UHFFFAOYSA-N
SMILES: [O-][N+]1=C2C=CC(Cl)=CC2=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results