accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1735-54-2

IUPAC Name: 4-chloro-3-(trifluoromethyl)benzonitrile
Synonyms: 4-chloro-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 205.56
Molecular Formula: C8H3ClF3N
InChi Key: VBPYHJRQZZCFCW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC=C1Cl)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results