accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17354-04-0

IUPAC Name: 2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
Synonyms: 2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
Molecular Weight (g/mol): 222.2
Molecular Formula: C10H10N2O4
InChi Key: PKDJSFDIQCAPSY-UHFFFAOYSA-N
SMILES: COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results