accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1736-56-7

IUPAC Name: 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
Synonyms: 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
Molecular Weight (g/mol): 202.13
Molecular Formula: C9H5F3O2
InChi Key: BGOMXTCPIUNFKR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)C=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results