accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1736-87-4

CH3H3CFNOO

CAS RN 1736-87-4

IUPAC Name: 1-fluoro-2,3-dimethyl-4-nitrobenzene
Synonyms: 1-fluoro-2,3-dimethyl-4-nitrobenzene
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: GLDMIZKOJPVEIV-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(=C1C)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)