accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 17362-17-3

IUPAC Name: 3-(4-hydroxyphenyl)propanenitrile
Synonyms: 3-(4-hydroxyphenyl)propanenitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: KDMJGLYRWRHKJS-UHFFFAOYSA-N
SMILES: OC1=CC=C(CCC#N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results