CAS RN 17364-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Palmitoyl-sn-glycero-3-phosphocholine, TRC
CAS: 17364-16-8 Molecular Formula: C24H50NO7P Molecular Weight (g/mol): 495.63 Synonym: (7R)-3,5,9-4,7-Dihydroxy-N,N,N-trimethyl-10-oxotrioxa-4-phosphapentacosan-1-aminium 4-Oxide Inner Salt,1-Hexadecanoyl-sn-glycerol-3-phosphorylcholine,1-Hexadecanoyllysolecithin,1-O-Palmitoyl-2-lyso-sn-glycero-3-phosphocholine,MPPC,Palmitoyl L-Lysophosphatidylcholine,Palmitoyl L-α-Lysolecithin,Palmitoyl L-α-Lysophosphatidylcholine,γ-Palmitoyl-L-α-lysophosphatidylcholine; SMILES: O=P([O-])(OCC[N+](C)(C)C)OC[C@H](O)COC(CCCCCCCCCCCCCCC)=O