accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OF

CAS RN 1737-19-5

IUPAC Name: 1-(3-fluorophenyl)propan-2-one
Synonyms: 1-(3-fluorophenyl)propan-2-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: UWCPYXSRCQVABG-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC(F)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results