accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHFFF

CAS RN 1737-26-4

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 1-[4-(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 190.17
Molecular Formula: C9H9F3O
InChi Key: YMXIDIAEXNLCFT-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results