accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17376-04-4

IUPAC Name: (2-iodoethyl)benzene
Synonyms: (2-iodoethyl)benzene
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: KVTHPKXDLVYNCH-UHFFFAOYSA-N
SMILES: ICCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results