CAS RN 17377-95-6
IUPAC Name:
N-benzyl-4-methoxyaniline
Synonyms:
N-benzyl-4-methoxyaniline
Molecular Weight (g/mol):
213.28
Molecular Formula:
C14H15NO
InChi Key:
LIJJGMDKVVOEFT-UHFFFAOYSA-N
SMILES:
COC1=CC=C(NCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
N-Benzyl-4-methoxyaniline, 99%, Thermo Scientific™
CAS: 17377-95-6 Molecular Formula: C14H15NO Molecular Weight (g/mol): 213.28 MDL Number: MFCD00059298 InChI Key: LIJJGMDKVVOEFT-UHFFFAOYSA-N Synonym: n-benzyl-p-anisidine,n-4-methoxyphenyl-n-benzylamine,4-methoxyphenyl benzylamine,n-benzyl-para-anisidine,acmc-1bvsm,n-benzyl-p-methoxyaniline,cambridge id 5107549,n-benzyl-4-methoxy-aniline,n-4-methoxyphenyl benzylamine,n-4-methoxyphenyl benzyl amine PubChem CID: 519413 IUPAC Name: N-benzyl-4-methoxyaniline SMILES: COC1=CC=C(NCC2=CC=CC=C2)C=C1