accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CNOO

CAS RN 174092-82-1

IUPAC Name: 6-isocyano-2,3-dihydro-1,4-benzodioxine
Synonyms: 6-isocyano-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: AAUQOWLQNFIDQL-UHFFFAOYSA-N
SMILES: [C-]#[N+]C1=CC2=C(OCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results