accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 174125-30-5

CH3SiH3CH3CCl

CAS RN 174125-30-5

IUPAC Name: (5-chloropent-1-yn-1-yl)triethylsilane
Synonyms: (5-chloropent-1-yn-1-yl)triethylsilane
Molecular Weight (g/mol): 216.82
Molecular Formula: C11H21ClSi
InChi Key: YOFYXFCOGXKCGG-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)C#CCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results