accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17452-27-6

SCNN

CAS RN 17452-27-6

IUPAC Name: 3-isothiocyanatopyridine
Synonyms: 3-isothiocyanatopyridine
Molecular Weight (g/mol): 136.17
Molecular Formula: C6H4N2S
InChi Key: VMSZFBSYWXMXRF-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results