accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1746-23-2

CH3H3CCH3CH2

CAS RN 1746-23-2

IUPAC Name: 1-tert-butyl-4-ethenylbenzene
Synonyms: 1-tert-butyl-4-ethenylbenzene
Molecular Weight (g/mol): 160.26
Molecular Formula: C12H16
InChi Key: QEDJMOONZLUIMC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results