accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBr

CAS RN 1746-28-7

IUPAC Name: 1-bromo-4-(2-bromoethyl)benzene
Synonyms: 1-bromo-4-(2-bromoethyl)benzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: APTDRDYSJZQPPI-UHFFFAOYSA-N
SMILES: BrCCC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)