accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17469-89-5

(S)OHONH3CCH3

CAS RN 17469-89-5

IUPAC Name: 2-(dimethylamino)-3-phenylpropanoic acid
Synonyms: 2-(dimethylamino)-3-phenylpropanoic acid
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: HOGIQTACRLIOHC-UHFFFAOYNA-N
SMILES: CN(C)C(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results