CAS RN 174899-66-2
IUPAC Name:
1-butyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms:
1-butyl-3-methylimidazolium triflate
Molecular Weight (g/mol):
288.29
Molecular Formula:
C9H15F3N2O3S
InChi Key:
FRZPYEHDSAQGAS-UHFFFAOYSA-M
SMILES:
[O-]S(=O)(=O)C(F)(F)F.CCCCN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1-n-Butyl-3-methylimidazolium trifluoromethanesulfonate, 98%
CAS: 174899-66-2 Molecular Formula: C9H15F3N2O3S Molecular Weight (g/mol): 288.285 MDL Number: MFCD03427620 InChI Key: FRZPYEHDSAQGAS-UHFFFAOYSA-M Synonym: 1-butyl-3-methylimidazolium trifluoromethanesulfonate,1-butyl-3-methylimidazolium trifluoromethansulfonate,1-butyl-3-methylimidazolium triflate,c4mim triflate,bmim otf,acmc-1bwln,basionics™ vs 12,dsstox_cid_29088,dsstox_rid_83307,dsstox_gsid_49232 PubChem CID: 2734246 IUPAC Name: 1-butyl-3-methylimidazol-3-ium;trifluoromethanesulfonate SMILES: CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]