accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNH2

CAS RN 1750-42-1

IUPAC Name: 1,2-oxazol-3-amine
Synonyms: 1,2-oxazol-3-amine
Molecular Weight (g/mol): 84.08
Molecular Formula: C3H4N2O
InChi Key: RHFWLPWDOYJEAL-UHFFFAOYSA-N
SMILES: NC1=NOC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results