accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOSOOClNOO

CAS RN 175135-00-9

IUPAC Name: 4-(2-chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride
Synonyms: 4-(2-chloro-6-nitrophenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 348.15
Molecular Formula: C12H7Cl2NO5S
InChi Key: CRTUVOFOPIFTQS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results