accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175135-09-8

IUPAC Name: 2,6-dibromo-3-chloro-4-fluoroaniline
Synonyms: 2,6-dibromo-3-chloro-4-fluoroaniline
Molecular Weight (g/mol): 303.35
Molecular Formula: C6H3Br2ClFN
InChi Key: PRZBEZGEEPNKRO-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C(Cl)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results