accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3ClOCH3

CAS RN 175136-01-3

IUPAC Name: methyl 2-chloro-3,4-dimethoxybenzoate
Synonyms: methyl 2-chloro-3,4-dimethoxybenzoate
Molecular Weight (g/mol): 230.64
Molecular Formula: C10H11ClO4
InChi Key: RYYAHUFRPCBHCT-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C(OC)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results