accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFPFFFFFFFFFFFFFFF

CAS RN 175136-62-6

IUPAC Name: tris[3,5-bis(trifluoromethyl)phenyl]phosphane
Synonyms: tris[3,5-bis(trifluoromethyl)phenyl]phosphane
Molecular Weight (g/mol): 670.28
Molecular Formula: C24H9F18P
InChi Key: ITJHLZVYLDBFOJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results