accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOSOHO

CAS RN 175136-92-2

IUPAC Name: 5-oxo-1-[(thiophen-2-yl)methyl]pyrrolidine-3-carboxylic acid
Synonyms: 5-oxo-1-(thiophen-2-ylmethyl)pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 225.26
Molecular Formula: C10H11NO3S
InChi Key: HNEDKCTVOYJYQI-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CC2=CC=CS2)C(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results