accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175137-11-8

IUPAC Name: methyl 3-chloro-4-methylthiophene-2-carboxylate
Synonyms: methyl 3-chloro-4-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: BUFSIDWTJVUAER-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C(C)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results