CAS RN 175137-20-9
IUPAC Name:
5-(4-fluorophenyl)-3-(methylsulfanyl)-1H-pyrazole
Synonyms:
3-(4-fluorophenyl)-5-(methylsulfanyl)-2H-pyrazole
Molecular Weight (g/mol):
208.25
Molecular Formula:
C10H9FN2S
InChi Key:
ZDFTZMJWTJLPRE-UHFFFAOYSA-N
SMILES:
CSC1=NNC(=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
3-(4-Fluorophenyl)-5-(methylthio)-1H-pyrazole, 97%, Thermo Scientific™
CAS: 175137-20-9 Molecular Formula: C10H9FN2S Molecular Weight (g/mol): 208.25 MDL Number: MFCD00052075 InChI Key: ZDFTZMJWTJLPRE-UHFFFAOYSA-N Synonym: 3-4-fluorophenyl-5-methylthio pyrazole,3-4-fluorophenyl-5-methylthio-1h-pyrazole,3-4-fluorophenyl-5-methylsulfanyl-1h-pyrazole,5-4-fluorophenyl-3-methylsulfanyl-1h-pyrazole,3-4-fluorophenyl-5-methylthiopyrazole,maybridge1_003702,5-4-fluorophenyl-3-methylthio-1h-pyrazole,3-4-fluoro-phenyl-5-methylsulfanyl-1h-pyrazole,3-4-fluorophenyl-5-methylsulfanyl-2h-pyrazole PubChem CID: 2737479 SMILES: CSC1=NNC(=C1)C1=CC=C(F)C=C1