accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175172-61-9

CH3OOSOOH3C

CAS RN 175172-61-9

IUPAC Name: 2-methoxyethyl methanesulfonate
Synonyms: 2-methoxyethyl methanesulfonate
Molecular Weight (g/mol): 154.18
Molecular Formula: C4H10O4S
InChi Key: BCKAHDGFNHDQST-UHFFFAOYSA-N
SMILES: COCCOS(C)(=O)=O
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)