accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1752-88-1

IUPAC Name: hydrogen pyrazolidin-3-one chloride
Synonyms: hydrogen pyrazolidin-3-one chloride
Molecular Weight (g/mol): 122.55
Molecular Formula: C3H7ClN2O
InChi Key: HAYWTXQRYXYRAF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=C1CCNN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results