accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOSSOOCH3NH2

CAS RN 175201-73-7

IUPAC Name: methyl 3-amino-4-methanesulfonylthiophene-2-carboxylate
Synonyms: methyl 3-amino-4-methanesulfonylthiophene-2-carboxylate
Molecular Weight (g/mol): 235.27
Molecular Formula: C7H9NO4S2
InChi Key: ZFMCGKFMQDQBLY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(=CS1)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results