accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONSHN

CAS RN 175202-77-4

IUPAC Name: 6-(4-methoxyphenyl)-1,2-dihydropyrimidine-2-thione
Synonyms: 4-(4-methoxyphenyl)-3H-pyrimidine-2-thione
Molecular Weight (g/mol): 218.27
Molecular Formula: C11H10N2OS
InChi Key: KSXDNCUHISGCTF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results